accessibility menu, dialog, popup

UserName

CAS RN 6106-21-4

IUPAC Name: disodium butanedioate hexahydrate
Synonyms: disodium succinate hexahydrate sodium succinate.6H2O disodium butanedioate.6H2O disodium succinate.6H2O succinic acid disodium salt hexahydrate sodium succinate hexahydrate
Molecular Weight (g/mol): 270.14
Molecular Formula: C4H16Na2O10
InChi Key: ZBTUYCUNQBRXOR-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O
Quantity 
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results