Learn More
Sodium succinate hexahydrate
Description
Sodium succinate hexahydrate is used as an ingredient of topical preparations for the treatment of cataract.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 120°C-6H2O |
| Linear Formula | NaO2CCH2CH2CO2Na·6H2O |
| Quantity | 250 g |
| Beilstein | 3921044 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,8679 |
| Solubility Information | It is soluble in water at a concentration of 270g/L at 20°C (68°F) . |
| Formula Weight | 270.14 (162.05 Anhydrous) |
| Physical Form | Crystalline or Granular Powder |
| Chemical Name or Material | Sodium succinate hexahydrate |
Safety and Handling
- Sodium succinate hexahydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: Succinate, disodium,hexahydrate
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.