missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid, disodium salt hexahydrate, 97%

Product Code. 10796581 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10796581 500 g 500g
10215970 100 g 100g
10708932 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10796581 Supplier Thermo Scientific Acros Supplier No. 158755000
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6106-21-4 | C4H16Na2O10 | 270.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-21-4
Molecular Formula C4H16Na2O10
Molecular Weight (g/mol) 270.142
InChI Key ZBTUYCUNQBRXOR-UHFFFAOYSA-L
Synonym disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
PubChem CID 3083938
ChEBI CHEBI:63686
IUPAC Name disodium;butanedioate;hexahydrate
SMILES C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]

Specifications

Melting Point 120.0°C
Color Brown to Yellow
pH 7 to 9 (0.4M soln.)
Loss on Drying 37.0 % to 41.0 % (90°C)
Infrared Spectrum Authentic
Assay Percent Range 0.97
Packaging Glass bottle
Quantity 500 g
Beilstein 02, 606
Merck Index 15, 8808
Solubility Information Solubility in water: 270g/L water (25°C). Other solubilities: insoluble in alcohol
Formula Weight 270.13
Percent Purity 0.97
Insoluble Matter 0.005% max.
Physical Form Liquid
Chemical Name or Material Succinic acid, disodium salt hexahydrate, 0.97
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.