accessibility menu, dialog, popup

UserName

CH3OOO

CAS RN 611-13-2

IUPAC Name: methyl furan-2-carboxylate
Synonyms: methyl furoate
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: HDJLSECJEQSPKW-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CO1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (6)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (4)
  • (1)