missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-furoate, 98+%

Product Code. 11467876
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467876 500 g 500g
11447876 25 g 25g
11457876 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467876 Supplier Thermo Scientific Alfa Aesar Supplier No. A14009.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 611-13-2 | C6H6O3 | 126.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-13-2
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00003236
InChI Key HDJLSECJEQSPKW-UHFFFAOYSA-N
Synonym methyl 2-furoate, methyl pyromucate, methyl 2-furancarboxylate, methyl furoate, 2-furancarboxylic acid, methyl ester, 2-methoxycarbonyl furan, pyromucic acid methyl ester, methyl 2-furylcarboxylate, 2-furoic acid, methyl ester, furoic acid, methyl ester
PubChem CID 11902
IUPAC Name methyl furan-2-carboxylate
SMILES COC(=O)C1=CC=CO1

Specifications

Density 1.179
Boiling Point 180°C to 181°C
Flash Point 73°C (163°F)
Refractive Index 1.487
Quantity 500 g
UN Number UN2810
Beilstein 111110
Merck Index 14,4307
Formula Weight 126.11
Percent Purity ≥98%
Chemical Name or Material Methyl 2-furoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.