accessibility menu, dialog, popup

UserName

OHN

CAS RN 611-20-1

IUPAC Name: 2-hydroxybenzonitrile
Synonyms: O-cyanophenol
Molecular Weight (g/mol): 119.12
Molecular Formula: C7H5NO
InChi Key: CHZCERSEMVWNHL-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C#N
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (3)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)