missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanophenol, 99%

Product Code. 10367530 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10367530 5 g Glass bottle 5g
10741241 25 g Glass bottle 25g
10022040 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10367530 Supplier Thermo Scientific Acros Supplier No. 161410050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 611-20-1 | C7H5NO | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-20-1
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.12
MDL Number MFCD00002145
InChI Key CHZCERSEMVWNHL-UHFFFAOYSA-N
Synonym 2-cyanophenol, o-cyanophenol, salicylonitrile, o-hydroxybenzonitrile, benzonitrile, 2-hydroxy, salicylnitrile, benzonitrile, hydroxy, benzonitrile, o-hydroxy, o-hydoxybenzonitrile, 2-hydroxy-benzonitrile
PubChem CID 11907
IUPAC Name 2-hydroxybenzonitrile
SMILES C1=CC=C(C(=C1)C#N)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 95.0°C to 98.0°C
Color White to Brown
Boiling Point 149.0°C (14.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NCC6H4OH
Quantity 5 g
Beilstein 10, 96
Solubility Information Solubility in water: soluble
Formula Weight 119.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanophenol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Cyanophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.