accessibility menu, dialog, popup

UserName

BrBrO

CAS RN 611-91-6

IUPAC Name: 2,3-dibromo-1,3-diphenylpropan-1-one
Synonyms: 2,3-dibromo-1,3-diphenylpropan-1-one
Molecular Weight (g/mol): 368.07
Molecular Formula: C15H12Br2O
InChi Key: LYAGBKGGYRLVTR-UHFFFAOYNA-N
SMILES: BrC(C(Br)C1=CC=CC=C1)C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results