missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromo-3-phenylpropiophenone, 98%, Thermo Scientific™

Product Code. 10641895 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10641895 5 g Glass bottle 5g
10016332 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10641895 Supplier Thermo Scientific Acros Supplier No. 375080050
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 611-91-6
Molecular Formula C15H12Br2O
Molecular Weight (g/mol) 368.07
MDL Number MFCD00017861
InChI Key LYAGBKGGYRLVTR-UHFFFAOYNA-N
PubChem CID 95342
IUPAC Name 2,3-dibromo-1,3-diphenylpropan-1-one
SMILES BrC(C(Br)C1=CC=CC=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 156°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH(Br)CH(Br)C(O)C6H5
Quantity 5 g
Synonym chalcone dibromide, 2,3-dibromo-1,3-diphenyl-1-propanone, benzalacetophenone dibromide, 2,3-dibromo-3-phenylpropiophenone, 1-propanone, 2,3-dibromo-1,3-diphenyl, propiophenone, 2,3-dibromo-3-phenyl, 1-propanone,2,3-dibromo-1,3-diphenyl, dibromodiphenylpropanone, benzal acetophenone dibromide, propiophenone,3-dibromo-3-phenyl
Solubility Information Solubility in water: negligible
Formula Weight 368.07
Percent Purity 98%
Chemical Name or Material 2, 3-Dibromo-3-phenylpropiophenone
compliance-icons
Product Identifier
  • 2,3-Dibromo-3-phenylpropiophenone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.