accessibility menu, dialog, popup

UserName

H3COO

CAS RN 611-94-9

IUPAC Name: (4-methoxyphenyl)(phenyl)methanone
Synonyms: (4-methoxyphenyl)(phenyl)methanone
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: SWFHGTMLYIBPPA-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results