missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzophenone, 97%

Product Code. 10295120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10295120 25 g Glass bottle 25g
10041120 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10295120 Supplier Thermo Scientific Acros Supplier No. 125740250
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 611-94-9 | C14H12O2 | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-94-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00008403
InChI Key SWFHGTMLYIBPPA-UHFFFAOYSA-N
Synonym 4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, p-methoxybenzophenone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, phenyl p-anisyl ketone, 4-benzoylanisole, 4-methoxyphenyl phenylmethanone, unii-i4xj07373m, 4-methoxyphenyl-phenylmethanone
PubChem CID 69146
IUPAC Name (4-methoxyphenyl)-phenylmethanone
SMILES COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 58°C to 63°C
Color Orange-Yellow to White
Boiling Point 354°C to 356°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4COC6H5
Quantity 25 g
Beilstein 08, 159
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol and ether
Formula Weight 212.25
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 4-Methoxybenzophenone, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.