accessibility menu, dialog, popup

UserName

CH3OOH3COH3COCH3

CAS RN 6110-37-8

IUPAC Name: methyl 2,4-dimethoxy-6-methylbenzoate
Synonyms: methyl 2,4-dimethoxy-6-methylbenzoate
Molecular Weight (g/mol): 210.23
Molecular Formula: C11H14O4
InChi Key: QCEODMDPMRZZDI-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(OC)C=C(OC)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results