CAS RN 612-37-3
CAS RN 612-37-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Methyluric Acid, TRC
CAS: 612-37-3 Molecular Formula: C6 H6 N4 O3 Molecular Weight (g/mol): 182.14 IUPAC Name: 7-methyl-3,9-dihydropurine-2,6,8-trione SMILES: CN1C(=O)NC2=C1C(=O)NC(=O)N2