accessibility menu, dialog, popup

UserName

H2OH2ONaNaNaOHOOOOOO

CAS RN 6132-04-3

IUPAC Name: trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
Synonyms: sodium citrate sodium citrate dihydrate sodium citrate hydrate
Molecular Weight (g/mol): 294.1
Molecular Formula: C6H9Na3O9
InChi Key: NLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILES: O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)