missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Molecular Biology Reagents
BIOPHARMA

Product Code. 15313591
Change view
Click to view available options
Chemical Name or Material:
Saline Sodium Citrate Buffer
TAE Buffer
Tris Borate EDTA Buffer
Tris Hydrochloride Buffer
pH:
7 ±0.1
7.5 ±0.1
8 ±0.1
8.3 ±0.1
8.4 ±0.1
Unit Size:
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Chemical Name or Material pH Unit Size
15313591 Saline Sodium Citrate Buffer 7 ±0.1 Pack of 6
15343711 TAE Buffer 8.3 ±0.1 Pack of 6
15383711 Tris Hydrochloride Buffer 7.5 ±0.1 Pack of 6
15353711 Tris Borate EDTA Buffer 8.4 ±0.1 Pack of 6
15393711 Tris Hydrochloride Buffer 8 ±0.1 Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15313591 Supplier Corning™ Supplier No. 46020CM
  • ONLINE EXCLUSIVE PRICE
    286.00 €
    203.34 € / Pack of 6
    Save 82.66 €
    28% Off
 Restricted Item
Estimated Shipment: 05-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in molecular biology, nucleic acid purification applications

  • Certificate of Analysis available
  • Product can be diluted to obtain the desired concentration

Order Info

Shipped at ambient temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Saline Sodium Citrate Buffer
CAS 6132-04-3, 7732-18-5
Concentration 20X
pH 7 ±0.1
Physical Form Liquid
Quantity 6 x 1 L
Solution Type Buffer
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.