accessibility menu, dialog, popup

UserName

NaCH3CH3H3CSOOO

CAS RN 6148-75-0

IUPAC Name: sodium 2,4,6-trimethylbenzene-1-sulfonate
Synonyms: sodium 2,4,6-trimethylbenzenesulfonate
Molecular Weight (g/mol): 222.23
Molecular Formula: C9H11NaO3S
InChi Key: AOJUNZYQOYSGHT-UHFFFAOYSA-M
SMILES: [Na+].CC1=CC(C)=C(C(C)=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results