missing translation for 'onlineSavingsMsg'
Learn More

Mesitylenesulfonic acid sodium salt hemihydrate, 98%

Product Code. 11429517
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429517 500 g 500g
11409517 25 g 25g
11419517 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429517 Supplier Thermo Scientific Alfa Aesar Supplier No. B22029.36
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6148-75-0 | C9H11NaO3S | 222.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6148-75-0
Molecular Formula C9H11NaO3S
Molecular Weight (g/mol) 222.23
MDL Number MFCD00067712
InChI Key AOJUNZYQOYSGHT-UHFFFAOYSA-M
Synonym sodium mesitylenesulfonate, benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt, sodium 2,4,6-trimethylbenzenesulfonate, sodium 2,4,6-trimethylbenzenesulphonate, mesitylenesulfonic acid sodium salt hemihydrate, mesitylenesulfonic acid sodium salt, benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt 1:1, 2,4,6-trimethylbenzenesulfonic acid sodium salt, mesitylene sodium sulfonate, acmc-1bbae
PubChem CID 23661859
IUPAC Name sodium;2,4,6-trimethylbenzenesulfonate
SMILES [Na+].CC1=CC(C)=C(C(C)=C1)S([O-])(=O)=O

Specifications

Melting Point >300°C
Quantity 500 g
Beilstein 5784990
Formula Weight 231.26 (222.25 Anhydrous)
Percent Purity 98%
Chemical Name or Material Mesitylenesulfonic acid sodium salt hemihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.