accessibility menu, dialog, popup

UserName

CH3SOOClNOO

CAS RN 616-83-1

IUPAC Name: 4-methyl-3-nitrobenzene-1-sulfonyl chloride
Synonyms: 4-methyl-3-nitrobenzenesulfonyl chloride
Molecular Weight (g/mol): 235.64
Molecular Formula: C7H6ClNO4S
InChi Key: OQFYBGANSUNUAO-UHFFFAOYSA-N
SMILES: CC1=C(C=C(C=C1)S(Cl)(=O)=O)[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results