missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrobenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals

Product Code. 11478777
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478777 5 g 5g
11488777 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478777 Supplier Thermo Scientific Alfa Aesar Supplier No. A19931.06
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 616-83-1 | Molecular Formula: C7H6ClNO4S | Molecular Weight: 235.638 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-83-1
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.638
MDL Number MFCD00129811
InChI Key OQFYBGANSUNUAO-UHFFFAOYSA-N
Synonym 4-methyl-3-nitrobenzene-1-sulfonyl chloride, 2-nitrotoluene-4-sulfonyl chloride, 4-methyl-3-nitro-benzenesulfonyl chloride, 4-methyl-3-nitrobenzenesulfonylchloride, chloro 4-methyl-3-nitrophenyl sulfone, 4-methyl-3-nitrobenzolsulfonylchlorid, pubchem5674, acmc-1awd3, 3-nitro-p-toluenesulfonyl chloride, 3-nitro-4-methyl phenylsulfonyl chloride
PubChem CID 560638
IUPAC Name 4-methyl-3-nitrobenzenesulfonyl chloride
SMILES CC1=C(C=C(C=C1)S(=O)(=O)Cl)[N+](=O)[O-]

Specifications

Melting Point 33°C to 36°C
Boiling Point 152°C to 154°C (1 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 2696481
Sensitivity Moisture sensitive
Formula Weight 235.65
Percent Purity 95%
Chemical Name or Material 4-Methyl-3-nitrobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.