CAS RN 61617-00-3
CAS RN 61617-00-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4,(5)-Methyl Mercapto Benzimidazole Zinc Salt, TRC
CAS: 61617-00-3 Molecular Formula: C16H14N4S2Zn Molecular Weight (g/mol): 391.82 Synonym: Antioxidant ZMMBI,Nocrac MMBZ,Rhenogran ZMMBI 50,Vanox ZMTI,Vulkanox ZMB 2,Vulkanox ZMBI,ZMMBI,ZMTI,Zinc 2-Mercaptotoluimidazole,Zinc Methylmercaptobenzimidazole IUPAC Name: bis[(4-methyl-1H-benzimidazol-2-yl)sulfanyl]zinc SMILES: Cc1cccc2[nH]c(S[Zn]Sc3nc4c(C)cccc4[nH]3)nc12