accessibility menu, dialog, popup

UserName

H3COOOHOH3C

CAS RN 617-05-0

IUPAC Name: ethyl 4-hydroxy-3-methoxybenzoate
Synonyms: ethyl vanillate
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: MWAYRGBWOVHDDZ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(O)C(OC)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results