missing translation for 'onlineSavingsMsg'
Learn More

Ethyl vanillate, 97%

Product Code. 11397595
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397595 10 g 10g
11307605 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397595 Supplier Thermo Scientific Alfa Aesar Supplier No. L05798.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 617-05-0 | C10H12O4 | 196.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-05-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00017269
InChI Key MWAYRGBWOVHDDZ-UHFFFAOYSA-N
Synonym ethyl vanillate, vanillic acid, ethyl ester, 4-hydroxy-3-methoxybenzoic acid ethyl ester, benzoic acid, 4-hydroxy-3-methoxy-, ethyl ester, unii-v38jk4z93o, 3-methoxy-4-hydroxybenzoic acid, ethyl ester, m-anisic acid, 4-hydroxy-, ethyl ester, vanilicacidethylester, ethyl 4-hydroxy-3-methoxy-benzoate, ethylvanillate
PubChem CID 12038
IUPAC Name ethyl 4-hydroxy-3-methoxybenzoate
SMILES CCOC(=O)C1=CC=C(O)C(OC)=C1

Specifications

Melting Point 42°C to 46°C
Boiling Point 293°C
Quantity 10 g
Beilstein 2100025
Formula Weight 196.2
Percent Purity 97%
Chemical Name or Material Ethyl vanillate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.