accessibility menu, dialog, popup

UserName

CAS RN 618-98-4

CH3OONOO

CAS RN 618-98-4

IUPAC Name: ethyl 3-nitrobenzoate
Synonyms: ethyl-M-nitrobenzoate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: MKBIJCPQTPFQKQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC(=C1)[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results