missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-nitrobenzoate, 98+%

Product Code. 11493334
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493334 100 g 100g
11483334 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493334 Supplier Thermo Scientific Alfa Aesar Supplier No. A10626.22
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 618-98-4 | C9H9NO4 | 195.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-98-4
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00014702
InChI Key MKBIJCPQTPFQKQ-UHFFFAOYSA-N
Synonym ethyl m-nitrobenzoate, benzoic acid, 3-nitro-, ethyl ester, ethyl-m-nitrobenzoate, m-nitrobenzoic acid, ethyl ester, benzoic acid, m-nitro-, ethyl ester, acmc-20akzc, 3-nitrobenzoic acid ethyl, benzoic acid, 3-nitro-,ethyl ester, benzoic acid, m-nitro-, ethyl ester 8ci, r---2-2-diphenylphosphino phenyl-4-phenyl-2-oxazoline
PubChem CID 69261
IUPAC Name ethyl 3-nitrobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 40°C to 43°C
Boiling Point 297°C to 298°C
Quantity 100 g
Beilstein 1912880
Merck Index 14,3832
Formula Weight 195.18
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.