CAS RN 62625-28-9
CAS RN 62625-28-9
IUPAC Name:
sodium 2-[(3,5-dibromo-4-hydroxyphenyl)(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzene-1-sulfonate
Synonyms:
sodium 2-[(3,5-dibromo-4-hydroxyphenyl)(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
Molecular Weight (g/mol):
691.94
Molecular Formula:
C19H9Br4NaO5S
InChi Key:
TWKDJMRHSLWRFI-UHFFFAOYSA-M
SMILES:
[Na+].OC1=C(Br)C=C(C=C1Br)C(=C1C=C(Br)C(=O)C(Br)=C1)C1=CC=CC=C1S([O-])(=O)=O
1
–
2
of
2
results
1