CAS RN 6280-80-4
IUPAC Name:
2-(2-formylphenoxy)acetic acid
Synonyms:
2-formylphenoxyacetic acid
Molecular Weight (g/mol):
180.16
Molecular Formula:
C9H8O4
InChi Key:
ANWMNLAAFDCKMT-UHFFFAOYSA-N
SMILES:
OC(=O)COC1=CC=CC=C1C=O
1
–
1
of
1
results
1
Alfa Aesar™ 2-Formylphenoxyacetic acid, 98%, Thermo Scientific™
CAS: 6280-80-4 Molecular Formula: C9H8O4 Molecular Weight (g/mol): 180.16 MDL Number: MFCD00003315 InChI Key: ANWMNLAAFDCKMT-UHFFFAOYSA-N Synonym: 2-formylphenoxyacetic acid,2-2-formylphenoxy acetic acid,2-formylphenoxy acetic acid,o-formylphenoxyacetic acid,acetic acid, 2-formylphenoxy,2-carboxymethoxybenzaldehyde,ortho-formylphenoxyacetic acid,acetic acid, o-formylphenoxy,acetic acid, 2-2-formylphenoxy,2-formylphenoxyaceticacid PubChem CID: 46533 IUPAC Name: 2-(2-formylphenoxy)acetic acid SMILES: OC(=O)COC1=CC=CC=C1C=O