missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Formylphenoxyacetic acid, 98%, Thermo Scientific™

Product Code. 11452038
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452038 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11452038 Supplier Alfa Aesar™ Supplier No. A14605.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6280-80-4
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00003315
InChI Key ANWMNLAAFDCKMT-UHFFFAOYSA-N
Synonym 2-formylphenoxyacetic acid, 2-2-formylphenoxy acetic acid, 2-formylphenoxy acetic acid, o-formylphenoxyacetic acid, acetic acid, 2-formylphenoxy, 2-carboxymethoxybenzaldehyde, ortho-formylphenoxyacetic acid, acetic acid, o-formylphenoxy, acetic acid, 2-2-formylphenoxy, 2-formylphenoxyaceticacid
PubChem CID 46533
IUPAC Name 2-(2-formylphenoxy)acetic acid
SMILES OC(=O)COC1=CC=CC=C1C=O

Specifications

Melting Point 131°C to 134°C
Quantity 25 g
Beilstein 2096797
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material 2-Formylphenoxyacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.