accessibility menu, dialog, popup

UserName

CAS RN 630-25-1

ClClBrClClBr

CAS RN 630-25-1

IUPAC Name: 1,2-dibromo-1,1,2,2-tetrachloroethane
Synonyms: 1,2-dibromo-1,1,2,2-tetrachloroethane
Molecular Weight (g/mol): 325.63
Molecular Formula: C2Br2Cl4
InChi Key: WJUKOGPNGRUXMG-UHFFFAOYSA-N
SMILES: ClC(Cl)(Br)C(Cl)(Cl)Br
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)