missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromotetrachloroethane, 97%

Product Code. 10637322 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10637322

Brand: Thermo Scientific Acros 158460250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 630-25-1 | C2Br2Cl4 | 325.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-25-1
Molecular Formula C2Br2Cl4
Molecular Weight (g/mol) 325.63
MDL Number MFCD00000780
InChI Key WJUKOGPNGRUXMG-UHFFFAOYSA-N
Synonym 1,2-dibromotetrachloroethane, dbtce, sym-dibromotetrachloroethane, ethane, 1,2-dibromo-1,1,2,2-tetrachloro, unii-0sgg217efr, 0sgg217efr, 1,2-dibrom-1,1,2,2-tetrachlorethan, brccl2ccl2br, 1,2-dibromotetrachloroethan, 1,2-dibrom-tetrachlorethane
PubChem CID 69426
IUPAC Name 1,2-dibromo-1,1,2,2-tetrachloroethane
SMILES C(C(Cl)(Cl)Br)(Cl)(Cl)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 220.0°C to 222.0°C
Color White to Beige
Density 2.7100g/mL
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrCCl2CCl2Br
Quantity 25 g
Beilstein 01, 93
Fieser 01,209
Specific Gravity 2.71
Solubility Information Solubility in water: insoluble
Formula Weight 325.63
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 1, 2-Dibromotetrachloroethane, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Dibromotetrachloroethane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.