accessibility menu, dialog, popup

UserName

SHBr

CAS RN 6320-01-0

IUPAC Name: 3-bromobenzene-1-thiol
Synonyms: 3-bromobenzenethiol
Molecular Weight (g/mol): 189.07
Molecular Formula: C6H5BrS
InChi Key: HNGQQUDFJDROPY-UHFFFAOYSA-N
SMILES: SC1=CC=CC(Br)=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)