missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophenol, 95%

Product Code. 10636364 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10636364 5 g Glass bottle 5g
10195890 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10636364 Supplier Thermo Scientific Acros Supplier No. 303710050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6320-01-0 | C6H5BrS | 189.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6320-01-0
Molecular Formula C6H5BrS
Molecular Weight (g/mol) 189.07
MDL Number MFCD00009603
InChI Key HNGQQUDFJDROPY-UHFFFAOYSA-N
Synonym 3-bromothiophenol, m-bromobenzenethiol, m-bromothiophenol, 3-bromo thiophenol, 3-bromobenzene-1-thiol, benzenethiol, 3-bromo, 3-bromophenyl mercaptan, 3-bromo-benzenethiol, pubchem6805, #
PubChem CID 80597
IUPAC Name 3-bromobenzenethiol
SMILES SC1=CC=CC(Br)=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White to Beige
Density 1.6320g/mL
Boiling Point 123°C to 124°C (40.0 mmHg)
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4SH
Refractive Index 1.6320 to 1.634
Quantity 5 g
Specific Gravity 1.632
Formula Weight 189.07
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromothiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.