accessibility menu, dialog, popup

UserName

H3COClNCO

CAS RN 63429-99-2

IUPAC Name: 4-chloro-2-isocyanato-1-methoxybenzene
Synonyms: 4-chloro-2-isocyanato-1-methoxybenzene
Molecular Weight (g/mol): 183.59
Molecular Formula: C8H6ClNO2
InChi Key: CDRZZFCLBUGMMB-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C=C1N=C=O
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results