missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2-methoxyphenyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10437632
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10437632 10 g 10g
10089521 1 g 1g
10148054 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10437632 Supplier Thermo Scientific Maybridge Supplier No. SB01657EA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 63429-99-2
Molecular Formula C8H6ClNOS
Molecular Weight (g/mol) 199.652
MDL Number MFCD00041073
InChI Key WGLDKQQXEWPFAR-UHFFFAOYSA-N
Synonym 5-chloro-2-methoxyphenyl isothiocyanate, 4-chloro-2-isothiocyanato-1-methoxy-benzene, 2-methoxy-5-chlorophenyl isothiocyanate, 5-chloro-2-methoxybenzenisothiocyanate, 4-chloranyl-2-isothiocyanato-1-methoxy-benzene, acmc-20am4k, 4-chloro-2-isothiocyanatoanisole, 5-chloro-2-methoxyphenylisothiocyanate, 5-chloro-2-methoxyphenyl-isothiocyanate, benzene, 4-chloro-2-isothiocyanato-1-methoxy
PubChem CID 737165
IUPAC Name 4-chloro-2-isothiocyanato-1-methoxybenzene
SMILES COC1=C(C=C(C=C1)Cl)N=C=S

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 199.66
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-methoxyphenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.