accessibility menu, dialog, popup

UserName

BrBrBrBr

CAS RN 636-28-2

IUPAC Name: 1,2,4,5-tetrabromobenzene
Synonyms: 1,2,4,5-tetrabromobenzene
Molecular Weight (g/mol): 393.7
Molecular Formula: C6H2Br4
InChi Key: QCKHVNQHBOGZER-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=C(Br)C=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)