missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrabromobenzene, 94%

Product Code. 11393127
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393127 10 g 10g
11303137 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393127 Supplier Thermo Scientific Alfa Aesar Supplier No. L13539.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 636-28-2 | C6H2Br4 | 393.70 g/mol

1,2,4,5-Tetrabromobenzene is used in the synthesis of diethynyltriptycene-linked dipyridyl ligands, in the generation of dibenzyne. It is also used to produce 6,7-dibromo-1,4-epoxy-1,4-dihydronaphthalene at temperature of -23°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4,5-Tetrabromobenzene is used in the synthesis of diethynyltriptycene-linked dipyridyl ligands, in the generation of dibenzyne. It is also used to produce 6,7-dibromo-1,4-epoxy-1,4-dihydronaphthalene at temperature of -23°C.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-28-2
Molecular Formula C6H2Br4
Molecular Weight (g/mol) 393.70
MDL Number MFCD00000063
InChI Key QCKHVNQHBOGZER-UHFFFAOYSA-N
Synonym benzene, 1,2,4,5-tetrabromo, 2,3,5,6-tetrabromobenzene, unii-m25dk66lcz, m25dk66lcz, pubchem4448, 1,4,5-tetrabromobenzene, 2,5,6-tetrabromobenzene, acmc-1b6z1, benzene,2,4,5-tetrabromo, benzene,1,2,4,5-tetrabromo
PubChem CID 12486
IUPAC Name 1,2,4,5-tetrabromobenzene
SMILES BrC1=CC(Br)=C(Br)C=C1Br

Specifications

Melting Point 175°C to 180°C
Quantity 10 g
Beilstein 1365830
Solubility Information Insoluble in water.
Formula Weight 393.7
Percent Purity 94%
Chemical Name or Material 1,2,4,5-Tetrabromobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.