accessibility menu, dialog, popup

UserName

H2NNH2

CAS RN 646-19-5

IUPAC Name: heptane-1,7-diamine
Synonyms: 1,7-diaminoheptane
Molecular Weight (g/mol): 130.24
Molecular Formula: C7H18N2
InChi Key: PWSKHLMYTZNYKO-UHFFFAOYSA-N
SMILES: NCCCCCCCN
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (4)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)