missing translation for 'onlineSavingsMsg'
Learn More

Heptane-1,7-diamine, TRC

Product Code. 17964185
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17964185 1 g 1g
17933215 250 mg 250mg
17960575 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17964185 Supplier Toronto Research Chemicals Supplier No. TRCH9986351G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-19-5
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.23
Synonym (7-Aminoheptyl)amine, 1,7-Diamino-n-heptane, 1,7-Diaminoheptane, 1,7-Heptamethylenediamine, 1,9-Diazanonane, Heptamethylenediamine, NSC 45777;alpha,ω-Heptanediamine;
IUPAC Name heptane-1,7-diamine
SMILES NCCCCCCCN

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H18N2/c8-6-4-2-1-3-5-7-9/h1-9H2
Formula Weight 130.147
Percent Purity >95%
Chemical Name or Material Heptane-1,7-diamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.