missing translation for 'onlineSavingsMsg'
Learn More

1,7-Diaminoheptane, 98%

Product Code. 11317875
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317875 25 g 25g
11307875 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317875 Supplier Thermo Scientific Alfa Aesar Supplier No. L02853.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 646-19-5 | C7H18N2 | 130.24 g/mol

1,7-Diaminoheptane is used in the preparation of 7-amino-1-guanidinooctane and 1,7-diamino-trans-hept-3-ene, which is used as a deoxyhypusine synthase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,7-Diaminoheptane is used in the preparation of 7-amino-1-guanidinooctane and 1,7-diamino-trans-hept-3-ene, which is used as a deoxyhypusine synthase inhibitor.

Solubility
Soluble in water, ethanol, acetone, ether and benzene.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-19-5
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.24
MDL Number MFCD00008246
InChI Key PWSKHLMYTZNYKO-UHFFFAOYSA-N
Synonym 1,7-diaminoheptane, 1,7-heptanediamine, heptamethylenediamine, chembl28242, h2n ch2 7nh2, heptan-1,7-diamin, 1,9-diazanonane, acmc-1atsg, 1,7-heptamethylenediamine
PubChem CID 69533
IUPAC Name heptane-1,7-diamine
SMILES NCCCCCCCN

Specifications

Melting Point 26°C to 30°C
Density 0.86
Boiling Point 223°C to 225°C
Flash Point 87°C (188°F)
Odor Amine-like
Linear Formula H2N(CH2)7NH2
Refractive Index 1.459
Quantity 25 g
UN Number UN3259
Beilstein 1734159
Sensitivity Air Sensitive
Solubility Information Soluble in water,ethanol,acetone,ether and benzene.
Formula Weight 130.24
Percent Purity 98%
Chemical Name or Material 1,7-Diaminoheptane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.