CAS RN 64902-72-3
IUPAC Name:
1-(2-chlorobenzenesulfonyl)-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)urea
Synonyms:
chlorsulfuron
Molecular Weight (g/mol):
357.77
Molecular Formula:
C12H12ClN5O4S
InChi Key:
VJYIFXVZLXQVHO-UHFFFAOYSA-N
SMILES:
COC1=NC(C)=NC(NC(=O)NS(=O)(=O)C2=CC=CC=C2Cl)=N1
1
–
2
of
2
results
2
Chlorosulfuron, TRC
CAS: 64902-72-3 Molecular Formula: C12 H12 Cl N5 O4 S Molecular Weight (g/mol): 357.77 Synonym: 2-Chloro-N-[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]benzenesulfonamide,Chlorsulfuron,DPX 4189,DPX-W 4189,Glean,Glean 20DF,Glean 75,Glean 75DF,Kartes,Khardin,Kortes,Luhuanglong,N-(2-Chlorophenyl)sulfonyl-N'-(4-methyl-6-methoxy-2-triazinyl)urea,Tuligen,W 4189 IUPAC Name: 1-(2-chlorophenyl)sulfonyl-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)urea SMILES: COc1nc(C)nc(NC(=O)NS(=O)(=O)c2ccccc2Cl)n1