missing translation for 'onlineSavingsMsg'
Learn More

Chlorosulfuron, TRC

Product Code. 17922885
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922885 250 mg 250mg
17954525 100 mg 100mg
17991265 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17922885 Supplier Toronto Research Chemicals Supplier No. TRCC384978250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64902-72-3
Molecular Formula C12 H12 Cl N5 O4 S
Molecular Weight (g/mol) 357.77
Synonym 2-Chloro-N-[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]benzenesulfonamide, Chlorsulfuron, DPX 4189, DPX-W 4189, Glean, Glean 20DF, Glean 75, Glean 75DF, Kartes, Khardin, Kortes, Luhuanglong, N-(2-Chlorophenyl)sulfonyl-N'-(4-methyl-6-methoxy-2-triazinyl)urea, Tuligen, W 4189
IUPAC Name 1-(2-chlorophenyl)sulfonyl-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)urea
SMILES COc1nc(C)nc(NC(=O)NS(=O)(=O)c2ccccc2Cl)n1

Specifications

Analyte or Component Names Chlorsulfuron
Shipping Condition Room Temperature
InChI Formula InChI=1S/C12H12ClN5O4S/c1-7-14-10(17-12(15-7)22-2)16-11(19)18-23(20,21)9-6-4-3-5-8(9)13/h3-6H,1-2H3,(H2,14,15,16,17,18,19)
Solution Type Neat
Industry Type Forensics
Quantity 250 mg
Formula Weight 357.0299
Percent Purity >95%
Chemical Name or Material Chlorsulfuron
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.