accessibility menu, dialog, popup

UserName

CAS RN 672310-11-1

IUPAC Name: (9H-fluoren-9-yl)methyl 3-oxopiperidine-1-carboxylate
Synonyms: 9H-fluoren-9-ylmethyl 3-oxopiperidine-1-carboxylate
Molecular Weight (g/mol): 321.38
Molecular Formula: C20H19NO3
InChi Key: ZHMRQZFSADRNCI-UHFFFAOYSA-N
SMILES: O=C(OCC1C2=CC=CC=C2C2=CC=CC=C12)N1CCCC(=O)C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results