missing translation for 'onlineSavingsMsg'
Learn More

1-Fmoc-3-piperidinone, 96%, Thermo Scientific™

Product Code. 15424958
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424958 250 mg 250mg
15404958 1 g 1g
15414958 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15424958 Supplier Thermo Scientific Alfa Aesar Supplier No. H58735.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 672310-11-1
Molecular Formula C20H19NO3
Molecular Weight (g/mol) 321.376
MDL Number MFCD03001740
InChI Key ZHMRQZFSADRNCI-UHFFFAOYSA-N
Synonym 1-n-fmoc-3-piperidone, 9h-fluoren-9-yl methyl 3-oxopiperidine-1-carboxylate, 1-fmoc-3-piperidone, 1-fmoc-3-piperidinone, n-fmoc-3-piperidinone, 1-fmoc-3-oxopiperidine, 9h-fluoren-9-yl methyl-3-oxopiperidine-1-carboxylate, 4 ? 3,4-difluoro-phenyl-4-propyl-bicyclohexyl, 9h-fluoren-9-yl methyl3-oxopiperidine-1-carboxylate
PubChem CID 1514170
IUPAC Name 9H-fluoren-9-ylmethyl 3-oxopiperidine-1-carboxylate
SMILES C1CC(=O)CN(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 90°C to 96°C
Quantity 250 mg
UN Number UN3077
Solubility Information Slightly soluble in water.
Formula Weight 321.38
Percent Purity 96%
Chemical Name or Material 1-Fmoc-3-piperidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.