accessibility menu, dialog, popup

UserName

CH3PClH3C

CAS RN 686-69-1

IUPAC Name: chlorodiethylphosphane
Synonyms: chlorodiethylphosphane
Molecular Weight (g/mol): 124.55
Molecular Formula: C4H10ClP
InChi Key: INJBDKCHQWVDGT-UHFFFAOYSA-N
SMILES: CCP(Cl)CC
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)