missing translation for 'onlineSavingsMsg'
Learn More

chloro(diethyl)phosphine, 95%

Product Code. 10255771 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10255771 1 g Glass bottle 1g
10206923 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10255771 Supplier Thermo Scientific Acros Supplier No. 350310010
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 686-69-1 | C4H10ClP | 124.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 686-69-1
Molecular Formula C4H10ClP
Molecular Weight (g/mol) 124.55
MDL Number MFCD00013621
InChI Key INJBDKCHQWVDGT-UHFFFAOYSA-N
Synonym chlorodiethylphosphine, diethylchlorophosphine, chloro diethyl phosphine, phosphinous chloride, diethyl, pubchem6480, chloro diethyl phosphane, acmc-1avkn, diethylphosphinous chloride, diethylphosphinous chloride #, chlorodiethyl phosphine
PubChem CID 69639
IUPAC Name chloro(diethyl)phosphane
SMILES CCP(CC)Cl

Specifications

Color Colorless to Yellow
Density 1.0230g/mL
Boiling Point 131.0°C to 132.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula (CH3CH2)2PCl
Refractive Index 1.4740 to 1.4760
Quantity 1 g
Specific Gravity 1.023
Formula Weight 124.55
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Chloro(diethyl)phosphine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloro(diethyl)phosphine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.