missing translation for 'onlineSavingsMsg'
Learn More

Chloro(diethyl)phosphine

Product Code. 15461167
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15461167 5 g 5g
15451167 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15461167 Supplier Thermo Scientific Alfa Aesar Supplier No. 030204.06
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 686-69-1 | C4H10ClP | 124.548 g/mol

Chloro(diethyl)phosphine is used as reagent. It is used as chemical and organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 686-69-1
Molecular Formula C4H10ClP
Molecular Weight (g/mol) 124.548
MDL Number MFCD00013621
InChI Key INJBDKCHQWVDGT-UHFFFAOYSA-N
Synonym chlorodiethylphosphine, diethylchlorophosphine, chloro diethyl phosphine, phosphinous chloride, diethyl, pubchem6480, chloro diethyl phosphane, acmc-1avkn, diethylphosphinous chloride, diethylphosphinous chloride #, chlorodiethyl phosphine
PubChem CID 69639
IUPAC Name chloro(diethyl)phosphane
SMILES CCP(CC)Cl

Specifications

Color Colorless to Yellow
Density 1.023 g/mL
Boiling Point 131°C to 132°C
Flash Point 18°C (66°F)
Linear Formula (C2H5)2PCl
Refractive Index 1.475
Quantity 5 g
UN Number UN2845
Sensitivity Air and moisture sensitive
Solubility Information Reacts violently.
Formula Weight 124.55
Physical Form Liquid
Chemical Name or Material Chloro(diethyl)phosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.