CAS RN 68986-76-5
IUPAC Name:
λ¹-copper(1+) thiophene-2-carboxylate
Synonyms:
λ¹-copper(1+) thiophene-2-carboxylate
Molecular Weight (g/mol):
190.68
Molecular Formula:
C5H3CuO2S
InChi Key:
SFJMFSWCBVEHBA-UHFFFAOYSA-M
SMILES:
[Cu+].[O-]C(=O)C1=CC=CS1
1
–
2
of
2
results
2
Copper(I) thiophene-2-carboxylate
CAS: 68986-76-5 Molecular Formula: C5H3CuO2S Molecular Weight (g/mol): 190.683 MDL Number: MFCD02183524 InChI Key: SFJMFSWCBVEHBA-UHFFFAOYSA-M Synonym: copper i thiophene-2-carboxylate,cutc,unii-7519wbl07l,copper i thiophenecarboxylate,copper 2-thienylcarboxylate,cuprous 2-thienylcarboxylate,copper 2-thiophenecarboxylate,copper 1+ thiophene-2-carboxylate,copper i thiophene-2-carboxylate mi PubChem CID: 11194830 IUPAC Name: copper(1+);thiophene-2-carboxylate SMILES: C1=CSC(=C1)C(=O)[O-].[Cu+]