accessibility menu, dialog, popup

UserName

CAS RN 69-22-7

H3CNNNCH3ONCH3OHOOOHOHOOHO

CAS RN 69-22-7

IUPAC Name: 1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione; 2-hydroxypropane-1,2,3-tricarboxylic acid
Synonyms: caffeine citrate
Molecular Weight (g/mol): 386.32
Molecular Formula: C14H18N4O9
InChi Key: RCQXSQPPHJPGOF-UHFFFAOYSA-N
SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O.CN1C=NC2=C1C(=O)N(C)C(=O)N2C
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)