accessibility menu, dialog, popup

UserName

H3CNHN

CAS RN 693-05-0

IUPAC Name: 3-(methylamino)propanenitrile
Synonyms: 3-(methylamino)propanenitrile
Molecular Weight (g/mol): 84.12
Molecular Formula: C4H8N2
InChi Key: UNIJBMUBHBAUET-UHFFFAOYSA-N
SMILES: CNCCC#N
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results