accessibility menu, dialog, popup

UserName

CAS RN 693288-97-0

ONOClHNOHO

CAS RN 693288-97-0

IUPAC Name: N-{4-chloro-2-[(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)methyl]phenyl}-2-hydroxybenzamide
Synonyms: N-{4-chloro-2-[(1,3-dioxoisoindol-2-yl)methyl]phenyl}-2-hydroxybenzamide
Molecular Weight (g/mol): 406.82
Molecular Formula: C22H15ClN2O4
InChi Key: UFOUABRZSDGGAZ-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C(=O)NC1=C(CN2C(=O)C3=CC=CC=C3C2=O)C=C(Cl)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)