missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPPHA, MedChemExpress
Description
CPPHA is potent and selective positive allosteric modulator (PAM) of the mGluR5 and mGluR1 (metabotropic glutamate receptor). CPPHA can potentiate responses of mGluR5 and mGluR1 to activation of these receptors. CPPHA is developed for the research of central nervous system disorders.
Specifications
Specifications
| Chemical Name or Material | CPPHA |
| CAS | 693288-97-0 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C22H15ClN2O4 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 100 mg/mL (245.81 mM; Need ultrasonic) |
| SMILES | O=C1N(CC2=CC(Cl)=CC=C2NC(C3=CC=CC=C3O)=O)C(C4=CC=CC=C41)=O |
| Molecular Weight (g/mol) | 406.82 |
| Formula Weight | 406.82 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?